Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RNF146 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11669R-A680
BOSSBS-11669R-A680EA 890 EUR InStock
BOSSBS-11669R-A680
Anti-RNF146 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. RNF146 (RING finger protein 146), also known as Dactylidin, is a 359 amino acid protein that contains one RING-type zinc finger and one WWE domain. Via its RING-type zinc finger, RNF146 may play a role in transcriptional regulation and protein degradation events. Defects in the gene encoding RNF146 are associated with Alzheimer's disease (AD) and may lead to a higher risk of breast cancer. Two isoforms of RNF146 exist due to alternative splicing events.

Type: Primary
Antigen: RNF146
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 95-160/359
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge