Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SETDB1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11670R-A488
BOSSBS-11670R-A488EA 876 EUR InStock
BOSSBS-11670R-A488
Anti-SETDB1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
ESET is a nuclear protein belonging to the histone-lysine methyltransferase family and to the Suvar3-9 subfamily. It is a highly conserved protein of 150 amino acids that has been implicated in chromatin structure modulation. ESET is excluded from cell nucleoli and areas of condensed chromatin and can associate with the nonpericentromeric regions of chromatin. The gene encoding for this protein, SETDB1, maps to chromosome 1q21. ESET is a histone methyltransferase, methylating Lys-9 of histone H3 and mutations within the SETDB1 gene abolishes its methyltransferase activity. This methylation acts as a tag for epigenetic transcriptional repression by rounding up HP1 proteins to methylated histones. ESET is widely expressed with highest levels found in testis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SETDB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge