Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CTNND2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11679R-CY5
BOSSBS-11679R-CY5EA 900 EUR InStock
BOSSBS-11679R-CY5
Anti-CTNND2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The Alpha-, Beta-, Gamma-, and Delta -catenins are proteins that bind to the highly conserved, intracellular cytoplasmic tail of E-cadherin. Together, the catenin/cadherin complexes play an important role mediating cellular adhesion. Alpha-catenin interacts with E-cadherin associated protein and also associates with other members of the cadherin family, such as N-cadherin and P-cadherin. Beta-catenin associates with the cytoplasmic portion of E-cadherin, which is necessary for the function of E-cadherin as an adhesion molecule. Beta-catenin also complexes with the tumor suppressor protein APC. Gamma-catenin, also known as plakoglobin, is a protein that binds ?catenin and N-cadherin. Gamma-catenin interacts with presenilin 1 and is expressed in the brain. The gene encoding Gamma-catenin maps to human chromosome 5p15.2. A hemizygous loss of the gene encoding Gamma-catenin leads to the mental retardation associated with Cri-du-Chat syndrome. In addition, the transmembrane phosphatase PTPm associates with catenin/ cadherin complexes and may regulate complex signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTNND2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge