Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-THOP1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11683R-CY7
BOSSBS-11683R-CY7EA 900 EUR InStock
BOSSBS-11683R-CY7
Anti-THOP1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Thimet oligopeptidase, also designated soluble metallo-endopeptidase, is a cytoplasmic protein belonging to the peptidase M3 family. The gene for the protein maps against chromosome 19q13.3. Thimet oligopeptidase can degrade the b-Amyloid precursor protein and generate amyloidogenic fragments. It is important in cytoplasmic peptide degradation and involved in metabolism of neuropeptides that are less than 20 amino acids in length. Thimet oligopeptidase is highly expressed in testis but can also be detected in liver, lung and kidney.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: THOP1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge