Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PCSK1N Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11687R-A647
BOSSBS-11687R-A647EA 1350 EUR InStock
BOSSBS-11687R-A647
Anti-PCSK1N Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
PCSK1N is a 260 amino acid protein that is both secreted and localized to the trans-Golgi network. Expressed in pancreas and brain, PCSK1N is thought to play a role in the control of the neuroendocrine secretory pathway and may also be involved in PCSK1 inhibition. The gene encoding PCSK1N maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes. In conjunction with chromosome Y, chromosome X is responsible for sex determination, as an X and a Y chromosome lead to normal male development, while two copies of an X chromosome lead to normal female development. There are a number of conditions related to an abnormal number and combination of sex chromosomes, some of which include Turner's syndrome, color blindness, hemophilia and Duchenne muscular dystrophy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PROSAAS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge