Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HAP40/F8a Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11696R-A350
BOSSBS-11696R-A350EA 893 EUR InStock
BOSSBS-11696R-A350
Anti-HAP40/F8a Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The Huntingtin protein contains a polyglutamine region, which leads to Huntingtin’s disease (HD) when the number of glutamine repeats exceeds thirty-five. The mutated Huntingtin protein acts within the nucleus to induce neurodegeneration by a cell-specific apoptotic mechanism. The loss of activity of the Huntingtin protein may be contributed to abnormal interactions between the mutant protein and other associated cellular proteins. Huntingtin interacts with a variety of proteins including HAP1, glyceraldehyde phosphate dehydrogenase (GAPDH), HAP40, Rab5 and HIP1. HAP40 mediates the recruitment of Huntingtin by Rab5 onto early endosomes. Specifically, this complex regulates endosome motility, which may be a key event of the pathogenetic process leading to neurodegeneration in HD.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HAP40/F8a
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge