Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HIPPI Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11697R-A555
BOSSBS-11697R-A555EA 893 EUR InStock
BOSSBS-11697R-A555
Anti-HIPPI Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Programmed neuronal cell death is a feature of neurodegenerative disorders such as Alzheimer's and Huntington's disease, which occur later in human life. Huntington’s disease at the molecular and cell level is characterized by polyglutamine expansion of the protein huntingtin (Htt) that leads to apoptotis-mediated neurodegenerative loss of medium spiny neurons throughout the striatum. Polyglutamine expansion reduces the level of association between Hip-1 and Htt, thereby increasing levels of free Hip-1 that then can be the candidate protein Hippi (Hip-1 protein interactor). The Hippi-Hip-1 heterodimer is a pro-apoptotic complex that recruits procaspase-8 and initiates caspase-8 activation, which may contribute to the neuronal cell death observed in individuals diagnosed with Huntington’s disease. The human hippi gene maps to chromosome 3q13.13 and encodes a 429 amino acid protein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HIPPI
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge