Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AIP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11705R-HRP
BOSSBS-11705R-HRPEA 900 EUR InStock
BOSSBS-11705R-HRP
Anti-AIP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
AIP1 (ASK-interacting protein 1), also known as DAB2IP (DAB2 interacting protein), is a 1,189 amino acid protein that localizes to both the membrane and the cytoplasm and contains one Ras-GAP domain, one PH domain and one C2 domain. Expressed at low levels in prostate tissue, AIP1 functions as a Ras GTPase-activating protein that interacts with ASK 1 and, via this interaction, disrupts the association of ASK 1 with an inhibitory 14-3-3 complex, thereby allowing free ASK 1 to function within the cell. AIP1 exists as multiple alternatively spliced isoforms and is down-regulated in prostate cancer tissue, suggesting a role in tumor suppression. Chromosomal aberrations in the gene encoding AIP1 are associated with acute myeloid leukemia (AML), implicating a role for AIP1 fusion proteins in tumorigenesis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: AIP1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge