Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ATP13A2 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11708R
BOSSBS-11708REA 725 EUR InStock
BOSSBS-11708R
Anti-ATP13A2 Rabbit Polyclonal Antibody
Antikörper
ATP13A2 is a 1,180 amino acid multi-pass membrane protein that belongs to the P5 subfamily of ATPases which play an important role in the transportation of inorganic cations. Expressed as multiple alternative spliced isoforms, ATP13A2 functions to catalyze the conversion of ATP to ADP and a free phosphate, thereby participating in the active transport of ions across cellular membranes. Defects in the gene encoding ATP13A2 are the cause of Kufor-Rakeb syndrome (KRS), a rare hereditary type of Parkinson’s disease that exhibits juvenile onset and is characterized by neurodegeneration and dementia. The ATP13A2 gene maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.

WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ATP13A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge