Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CLN8 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11715R-A350
BOSSBS-11715R-A350EA 893 EUR InStock
BOSSBS-11715R-A350
Anti-CLN8 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
CLN8, a 286 amino acid transmembrane protein, localizes mainly to the endoplasmic reticulum, but also partially to the ER-Golgi intermediate compartment (ERGIC). Mutations in the CLN8 gene cause neuronal ceroid lipofuscinosis 8 and progressive epilepsy with mental retardation (EPMR). Both disorders are forms of neuronal ceroid-lipofuscinose (NCL), a group of progressive neurodegenerative diseases found in children, characterized by failure of psychomotor development, impaired vision, seizures and premature death. The CLN8 protein is one of eight proteins in the CLN family, including CLN1-CLN7, which are associated with NCL.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLN8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge