Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CRBN Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11716R-A350
BOSSBS-11716R-A350EA 893 EUR InStock
BOSSBS-11716R-A350
Anti-CRBN Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
CRBN is a 442 amino acid protein which is highly concentrated in human brain tissue. CRBN functions are thought to be related to energy metabolism, learning and memory. Localized to the cytoplasm, CRBN acts as a protease in mitochondria and is thought to regulate the assembly of KCNT1, as well as the surface expression of KCNT1 in brain regions known to affect memory and learning, such as the hippocampus. The gene encoding CRBN belongs to a family of ATP-dependent lon proteases that play a role in membrane trafficking and proteolysis. Defects in the CRBN gene are associated with mild mental retardation.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRBN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge