Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HEXA Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11726R-A647
BOSSBS-11726R-A647EA 1350 EUR InStock
BOSSBS-11726R-A647
Anti-HEXA Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Hexosaminidase A (HEXA), also designated beta-Hexosaminidase A, is a trimer composed of one å chain, one ∫-A chain and one ∫-B chain and is found in the lysosomes of cells. HEXA, along with the cofactor CM2 activator protein, catalyzes the degradation of GM2 ganglioside and other molecules containing terminal N-acetyl hexosamines in the brain and other tissues. A mutation in the å subunit of hexosaminidase is the cause of Tay-Sachs disease (TSD), also known as GM2-gangliosidosis type I. TSD is a fatal autosomal recessive lysosomal storage disease of the central nervous system (CNS) caused by insufficient activity of the HEXA enzyme that results in a failure to process GM2 gangliosides. The accumulation of GM2 ganglioside in the absence of HEXA activity causes progressive destruction of the CNS.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HEXA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge