Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Doppel/DPL Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11732R
BOSSBS-11732REA 725 EUR InStock
BOSSBS-11732R
Anti-Doppel/DPL Rabbit Polyclonal Antibody
Antikörper
Prion diseases or transmissible spongiform encephalopathies (TSEs) are manifested as genetic, infectious or sporadic, lethal neurodegenerative disorders involving alterations of the prion protein (PrP). Infectious PrPSc is highly expressed in the brain of animals affected by TSEs, including scrapie in sheep, BSE in cattle, and Cruetzfeldt-Jacob disease in humans. The PRND gene locus, located on human chromosome 20p, encodes for the doppel protein (Dpl), which exhibits approximately 25% sequence homology with PrP. Dpl is characterized by an alpha-helical conformation, intramolecular disulfide bonds, and two N-linked oligosaccharides, and it is presented on the cell surface by a glycosylphosphatidylinositol anchor. Dpl is highly expressed in adult testis and heart and is detectable in the brain of neonatal mice. Dpl does not appear to contribute to prion disease progression, but ectopic expression of Dpl is implicated in neuronal degeneration of ataxic PRP-deficient mice. Dpl is also thought to play a role in angiogenesis, specifically maturation of the blood-brain barrier.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Doppel/DPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge