Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NPC2 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11736R
BOSSBS-11736REA 725 EUR InStock
BOSSBS-11736R
Anti-NPC2 Rabbit Polyclonal Antibody
Antikörper
NPC2 is a secreted protein mapping against gene 14q24.3 (1,2). NPC2 regulates the lipid composition of sperm membranes during maturation in the epididymis(1,2). Mutations in the NPC2 gene may cause Nieman-Pick type C2 disease and frontal lobe atrophy (1,2,3). Nieman-Pick type C2 is a fatal hereditary disease characterized by defective lysosome release of cholesterol (3). The disease is caused by HE1 deficiency, a lysosmal protein proven to be undetectable in fibroblasts from NPC2 patients (3). This differentiates NPC2 from NPC1, as NPC1 has HE1 protein present (3).

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NPC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge