Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DMPK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11742R-HRP
BOSSBS-11742R-HRPEA 900 EUR InStock
BOSSBS-11742R-HRP
Anti-DMPK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Myotonic dystrophy protein kinase is a multi-domain protein kinase found in muscle that is activated in response to G protein second messengers and proteolysis (1). DMPK is implicated in myotonic muscular dystrophy (DM), an autosomal dominant-inherited disorder that predominately affects skeletal and cardiac muscle and causes defects in cardiac conduction (2,3). DM arises through expansion of CTG repeats in the 3’-UTR of the DMPK gene (4). Mutant DMPK transcripts with an extended region of CUG repeats are retained in the nucleus (5). These transcripts also influence the expression of the DM locus-associated homeodomain protein (DMAHP)/SIX5, to mediate in part the DM phenotype (6). Other substrates for DMPK include myogenin, L-type calcium channels, and Phospholemman (PLM) (1).

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: DMPK
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge