Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PQBP1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11751R
BOSSBS-11751REA 725 EUR InStock
BOSSBS-11751R
Anti-PQBP1 Rabbit Polyclonal Antibody
Antikörper
Polyglutamine(Q) tract binding protein-1 (PQBP-1) is a transcription repressor that associates with polyglutamine tract-containing transcription regulators and causative genes for neurodegenerative disorders. Hepta- and di-amino acid repeat sequences rich in polar residues are essential for PQBP-1 to interact with polyglutamine tract-containing proteins (i.e. huntingtin, androgen receptor and Brain-2). PQBP-1 contains a WWP/WW domain that binds proline-rich motifs and a C2 domain that can influence Ca2+-dependent phospholipid signaling. PQBP-1 localizes to the nucleus and is present in neurons throughout the brain, with abundant levels in hippocampus, cerebellar cortex and olfactory bulb. The human PQBP-1 gene maps to chromosome Xp11.23.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: PQBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge