Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SGSH Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11756R-FITC
BOSSBS-11756R-FITCEA 900 EUR InStock
BOSSBS-11756R-FITC
Anti-SGSH Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Sulfatases are enzymes that hydrolyse a diverse range of sulfate esters. Deficiency of lysosomal sulfatases leads to human diseases characterized by the accumulation of either GAGs (glycosaminoglycans) or sulfolipids. Sulfamidase, also known as HSS, SFMD, MPS3A or SGSH, is a 502 amino acid lysosome that belongs to the sulfatase family. It has been suggested that sulfamidase may be involved in the lysosomal degradation of heparan sulfate. Defects in the gene encoding sulfamidase are the cause of Sanfilippo syndrome A, an autosomal recessive lysosomal storage disease caused by impaired degradation of heparan sulfate. Sanfilippo syndrome A is characterized by severe central nervous system degeneration but relatively mild somatic manifestations.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SGSH
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge