Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SULT4A1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11763R-A555
BOSSBS-11763R-A555EA 893 EUR InStock
BOSSBS-11763R-A555
Anti-SULT4A1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Sulfation is an essential conjugation reaction that increases the water solubility of many compounds, thereby influencing their renal excretion and also resulting in the formation of active metabolites. SULT4A1 (sulfotransferase family 4A, member 1), whose alternative names include brain sulfotransferase-like protein, nervous system sulfotransferase, NST, SULTX3, hBR-STL-1, BRSTL1, BR-STL-1, MGC40032 and DJ388M5.3, is a 284 amino acid protein showing cytoplasmic localization. As a member of the sulfotransferase 1 family, SULT4A1 plays a role in elimination of xenobiotics, activation of procarcinogens and regulation of hormones. SULT4A1 is highly expressed in cerebral cortex and frontal lobe, with lower expression in cerebellum, temporal and occipital lobes. Two SULT4A1 isoforms exist to alternative splicing events. The gene encoding SULT4A1 maps to human chromosome 22q13.2, a region which has been implicated in predisposition to schizophrenia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SULT4A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge