Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TIMM8A Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11769R-A555
BOSSBS-11769R-A555EA 893 EUR InStock
BOSSBS-11769R-A555
Anti-TIMM8A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The majority of mitochondrial-directed proteins are encoded by the nuclear genome and are transported to the mitochondria via regulated processes involving the mitochondrial Tom and Tim proteins (1). The mitochondrial Tim protein family is comprised of a large group of evolutionarily conserved proteins that are found in most eukaryotes (1,2). Import of nuclear-encoded precursor proteins into and across the mitochondrial inner membrane is mediated by two distinct complexes, the Tim23 complex and the Tim22 complex, which differ in their substrate specificity (1). Defects in Tim proteins are implicated in several neuro-degenerative diseases, suggesting important roles for Tim proteins in development and health (3,4). Tim8A and Tim8B, which map to human chromosomes Xq22.1 and 11q23.1-q23.2, respectively, are conserved proteins of the mitochondrial intermembrane space, which are organized in hetero-oligomeric complex with Tim13 (5,6,7). Tim8A is highly expressed in fetal and adult brain (5). Tim8A is mutated in deafness dystonia syndrome, a novel type of disease that causes severe neurological defects, thought to be caused by a defective mitochondrial protein transport system (5,8).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TIMM8A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge