Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TGase6/Transglutaminase 6 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11773R-A750
BOSSBS-11773R-A750EA 890 EUR InStock
BOSSBS-11773R-A750
Anti-TGase6/Transglutaminase 6 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antikörper
Terminally differentiating mammalian epidermal cells acquire an insoluble, 10 to 20 nm thick protein deposit on the intracellular surface of the plasma membrane, known as the cross-linked cell envelope (CE). The CE is a component of the epidermis that is generated through the formation of disulfide bonds and gamma-glutamyl-lysine isodipeptide bonds, which are formed by the action of transglutaminases (TGases). TGases are Ca2+-dependent enzymes, which catalyse the formation of isopeptide bonds by transferring an amine to a glutaminyl residue, thereby cross-linking glutamine residues and lysine residues in substrate proteins. TGases influence numerous biological processes, including blood coagulation, epidermal differentiation, seminal fluid coagulation, fertilisation, cell differentiation and apoptosis. TGase6 (transglutaminase 6), also known as TGM6, TGY or TGM3L, is a 706 amino acid protein that catalyses the cross-linking of proteins and the conjugation of proteins to polyamines. As a result of alternative splicing, two TGase6 isoforms exist.

Type: Primary
Antigen: TGase6/Transglutaminase 6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 551-640/706
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge