Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZFYVE27 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11777R-CY3
BOSSBS-11777R-CY3EA 900 EUR InStock
BOSSBS-11777R-CY3
Anti-ZFYVE27 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Zinc finger FYVE domain-containing protein 27 (ZFYVE27), also known as SPG33, is a 411 amino acid member of the FYVE-finger family of proteins. The FYVE domain is a cysteine-rich domain of about 70 amino acids that plays a role in the endosomal localization of the FYVE-finger proteins, and a majority of these proteins serve as regulators of endocytic membrane trafficking. ZFYVE27, a multi-pass membrane protein, is an endosomal protein that binds to Spastin, a protein that is primarily involved in microtubule dynamics and severing, vesicular trafficking and endosomal trafficking. Mutations in the gene encoding ZFTVE27 affect neuronal intracellular trafficking in the corticospinal tract and are thought to lead to hereditary spastic paraplegia (HSP), a neurodegenerative disorder, characterized by progressive paralysis of the legs, which is caused by impaired axonal transport. Five isoforms of ZFYVE27 exist as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZFYVE27
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge