Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SPG21 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11785R-A555
BOSSBS-11785R-A555EA 893 EUR InStock
BOSSBS-11785R-A555
Anti-SPG21 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Maspardin is a 308 amino acid cytoplasmic protein that is widely expressed. Belonging to the AB hydrolase superfamily, Maspardin colocalizes with CD4 on endosomal/trans-Golgi network. It is thought that Maspardin may act as a negative regulatory factor in CD4-dependent T-cell activation. Defects in the gene encoding Maspardin are the result of hereditary spastic paraplegia autosomal recessive type 21 (also designated Mast syndrome), an autosomal recessive neurodegenerative disorder characterized by a slow, gradual, progressive weakness and spasticity of the lower limbs. The gene encoding Maspardin is encoded by human chromosome 15, which houses over 700 genes and comprises nearly 3% of the human genome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPG21
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge