Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ENSA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11793R-HRP
BOSSBS-11793R-HRPEA 900 EUR InStock
BOSSBS-11793R-HRP
Anti-ENSA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Sulfonylureas are a class of drugs commonly used in the management of noninsulin-dependent diabetes mellitus. ENSA/ARPP-19, also known as ARPP-19e, is a 121 amino acid protein that localizes to the cytoplasm and belongs to the endosulfine family. ENSA/ARPP-19 is an endogenous regulator of the β-cell K(ATP) channels and reduces K(ATP) channel currents by inhibiting sulfonylurea from binding to the receptor thereby stimulating insulin secretion by pancreatic β cells. It is suggested that ENSA/ARPP-19 expression is decreased in brains of both Alzheimer's disease (AD) and Down's syndrome patients. Widely expressed with high levels in skeletal muscle and brain and lower levels in pancreas, ENSA/ARPP-19 is phosphorylated by PKA and exists as eight alternatively spliced isoforms. ENSA/ARPP-19 is encoded by a gene located on human chromosome 1q21.3.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ENSA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge