Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TMEM18 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11799R-FITC
BOSSBS-11799R-FITCEA 900 EUR InStock
BOSSBS-11799R-FITC
Anti-TMEM18 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
TMEM18 is a 140 amino acid multi-pass membrane protein that localizes to the nuclear membrane and is expressed in the brain. TMEM18 functions as a cell migration modulator which enhances the glioma-specific migration ability of neural precursor and neural stem cells.Overexpression of TMEM18 increases migration of human and murine neural stem cells, whereas knockdown of TMEM18 mRNA reduces cellular migration. Two specific single nucleotide polymorphisms (SNPs) within the TMEM18 gene locus known as rs6548238 and rs756131 have been linked to obesity susceptibility.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMEM18
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge