Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BRWD3 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11811R-CY3
BOSSBS-11811R-CY3EA 893 EUR InStock
BOSSBS-11811R-CY3
Anti-BRWD3 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Members of the WD repeat protein family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis and gene regulation. BRWD3 (bromodomain and WD repeat-containing protein 3) is a 1,802 amino acid protein expressed in fetal liver and most adult tissues. Existing as five alternatively spliced isoforms, BRWD3 contains two bromo domains, nine WD repeats and is thought to play a role in transcription by modifying chromatin. Mutations in the gene encoding BRWD3 are the cause of mental retardation X-linked type 93 (MRX93), which is also known as mental retardation X-linked with macrocephaly (XLMR). MRX93 is characterized by mild intellectual disability, macrocephaly, a prominent forehead and large cupped ears.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BRWD3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge