Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KALRN Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11861R-CY3
BOSSBS-11861R-CY3EA 900 EUR InStock
BOSSBS-11861R-CY3
Anti-KALRN Rabbit Polyclonal Antibody (Cy3®)
Antikörper
HAP1 (huntingtin-associated protein 1) binds to huntingtin (1). Huntingtin is a protein that contains a polyglutamine region and when the number of glutamine repeats exceeds 35, the gene encodes a version of huntingtin that leads to Huntington’s disease (HD) (2,3). The ability of HAP1 to bind to huntingtin is enhanced by an expanded polyglutamine repeat region (1). HAP1 shows neuronal localization and moves with huntingtin in nerve fibers (4,5). HAP1 is primarily expressed in brain tissue, with greater expression in the olfactory bulb and brain stem (1). Mouse HAP1 is localized to membrane-bound organelles including large endosomes, tubulovesicular structures and budding vesicles in neurons (6). Duo, also designated huntingtin-associated protein interacting protein or HAPIP, binds Huntingtin-associated protein 1 (HAP1) and may have a role in vesicle trafficking and cytoskeletal function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KALRN
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge