Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SHTN1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11863R-CY5
BOSSBS-11863R-CY5EA 900 EUR InStock
BOSSBS-11863R-CY5
Anti-SHTN1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Shootin1 is a 631 amino acid protein that belongs to the shootin family. The shootin1 protein contains three coiled-coil domains, a proline-rich region and interacts with RUFY3. Shootin1 is involved in the generation of internal asymmetric signals required for neuronal polarization. The shootin1 protein acts upstream of PI3K (phosphoinositide 3-kinase), by being required for spatially localized PI3K activity. By accumulating asymmetrically in a single neurite before polarization, shootin1 leads to axon induction for polarization, additionally the absence of shootin1 from the nascent axon's siblings by competition prevents the formation of surplus axons. Existing as seven alternatively spliced isoforms, the shootin1 gene is conserved in chimpanzee, dog, mouse, rat, chicken and zebrafish, and maps to human chromosome 10q25.3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Shootin/KIAA1598
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge