Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KIRREL3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11864R-A488
BOSSBS-11864R-A488EA 893 EUR InStock
BOSSBS-11864R-A488
Anti-KIRREL3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
NEPH2 is a 778 amino acid single-pass type I membrane protein that belongs to the nephrin-like protein family and immunoglobulin superfamily. Expressed in both fetal and adult brain, as well as podocytes of kidney glomeruli, NEPH2 contains five Ig-like C2-type (immunoglobulin-like) domains and is thought to plaly a role in the hematopoetic supportive capacity of stroma cells. NEPH2 undergoes alternative splicing to produce two isoforms and contains a C-terminal cytoplasmic domain which it uses to interact with Podocin, a podocyte protein involved in ultrafiltration. Defects in the gene encoding NEPH2 are associated with mental retardation autosomal dominant type 4 (MRD4).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIRREL3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge