Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LECT2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11869R-CY5
BOSSBS-11869R-CY5EA 893 EUR InStock
BOSSBS-11869R-CY5
Anti-LECT2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Leukocyte cell-derived chemotaxin 2 (LECT2) is a secreted protein with a neutrophil chemotactic activity. LECT2 is highly expressed in liver and shows diffuse immunostaining within the cytoplasm of hepatocytes. The LECT2 protein consists of 133 amino acids and three intramolecular disulfide bonds, and homologues of LECT2 have been widely identified in many vertebrates. LECT2 has a multifunctional role that extends from cell growth, differentiation, damage/repair process and carcinogenesis to autoimmune diseases. LECT2 expression is specifically induced in liver by β-catenin signaling. Serum LECT2 levels have been shown to increase in response to liver recovery, suggesting LECT2 may be used as a prognostic indicator.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LECT2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge