Anti-LHX5 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-11870R-CY5
BOSSBS-11870R-CY5EA 900 EUR InStock
BOSSBS-11870R-CY5
Anti-LHX5 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
During development, genetically distinct subtypes of motor neurons express unique combinations of LIM-type homeodomain factors, which regulate cell migration and guide motor axons to establish the fidelity of a binary choice in axonal trajectory. The LIM gene family encodes a set of gene products, which carry the LIM domain, a unique cysteine-rich zinc-binding domain. At least 40 members of this family have been identified in vertebrates and invertebrates, and are distributed into 4 groups according to the number of LIM domains and to the presence of homeodomains and kinase domains. The human LHX5 gene maps to chromosome 12q23-q24 and encodes a 402 amino acid protein. The hippocampus contains the neural circuitry, which is crucial for cognitive functions such as learning and memory. LHX5 regulates precursor cell proliferation and neuronal differentiation and migration during hippocampal development.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LHX5
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge