Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LINGO4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11873R-A750
BOSSBS-11873R-A750EA 890 EUR InStock
BOSSBS-11873R-A750
Anti-LINGO4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Lingo-4 is a 593 amino acid single-pass type I membrane protein that contains eleven LRR (leucine-rich) repeats, one Ig-like C2-type (immunoglobulin-like) domain, one LRRCT domain and one LRRNT domain. The gene that encodes Lingo-4 consists of approximately 5891 bases and maps to human chromosome 1q21.3. Comprising nearly 8% of the human genome, chromosome 1 spans 260 million base pairs, contains over 3000 genes and houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson's disease, Gaucher disease, schizophrenia and Usher syndrome.

Type: Primary
Antigen: LINGO4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 331-450/593
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge