Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LPPR4 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11875R
BOSSBS-11875REA 725 EUR InStock
BOSSBS-11875R
Anti-LPPR4 Rabbit Polyclonal Antibody
Antikörper
Phosphatidate phosphatases are a family of integral membrane glycoproteins that dephosphorylate a variety of lipid phosphates and play a role in signal transduction via the phospholipase D pathway. PAP-2 proteins function independently of Mg2+ and are insensitive to NEM (N-ethylmaleimide) inhibition. The lipid phosphates degraded by this family include ceramide 1-phosphate (C1P), sphingosine 1-phosphate (S1P), phosphatidic acid (PA) and lysophosphatidic acid (LPA). LPPR4 (lipid phosphate phosphatase-related protein type 4), also known as LPR4, PHP1, PRG1 or PRG-1, is a 763 amino acid multi-pass membrane protein that belongs to the PA-phosphatase related phosphoesterase family. Exclusively expressed in neurons, LPPR4 hydrolyzes lysophosphatidic acid (LPA) and facilitates axonal outgrowth during development and regenerative sprouting. LPPR4 exists as two alternatively spliced isoforms and is encoded by a gene located on human chromosome 1p21.2.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: LPPR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge