Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LRRTM2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11877R-CY7
BOSSBS-11877R-CY7EA 900 EUR InStock
BOSSBS-11877R-CY7
Anti-LRRTM2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The leucine-rich (LRR) repeat is a 20-30 amino acid motif that forms a hydrophobic Alpha/Beta horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. The LRRTM protein family plays a role in the regulation of various cellular events during nervous system development. Localizing predominantly to the nervous system, LRRTM family members are known to exhibit synaptogenic activity. LRRTM2 (leucine rich repeat transmembrane neuronal 2), also known as LRRN2, is a 516 amino acid single-pass type I membrane protein involved in the development maintenance of the vertebrate nervous system. Expressed in kidney and neuronal tissues, LRRTM2 contains ten LRR repeats and belongs to the LRRTM family. LRRTM2 is encoded by a gene that maps to human chromosome 5q31.2.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRRTM2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge