Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LGI3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11879R-A680
BOSSBS-11879R-A680EA 890 EUR InStock
BOSSBS-11879R-A680
Anti-LGI3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The leucine-rich (LRR) repeat is a 20 to 30 amino acid motif that forms a hydrophobic å/ horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. Leucine-rich glioma-inactivated protein 3 (LGI3), also known as LGI1-like protein 4 (LGIL4) or leucine-rich repeat LGI family member 3, is a 548 amino acid secretory protein. LGI3 contains five LRR repeats and seven EAR repeats. Widely expressed, with highest levels in brain and lung, LGI3 has been shown to colocalize with endocytosis-associated proteins, lipid raft markers, and Syntaxin. The gene encoding LGI3 maps to chromosome 8p21.3.

Type: Primary
Antigen: LGI3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-220/548
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge