Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MATH2/NEUROD6 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11884R-CY5
BOSSBS-11884R-CY5EA 900 EUR InStock
BOSSBS-11884R-CY5
Anti-MATH2/NEUROD6 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The Drosophila atonal gene produces a protein with basic helix loop helix (bHLH) domains that plays an essential role in the development of the Drosophila nervous system. Mammalian atonal homolog 2 (MATH-2) is a helix-loop-helix (HLH) transcription factor that is structurally homologous to the product of Drosophila atonal gene. MATH-2 is a 337 amino acid protein with an atonal-related basic HLH domain. In mice, expression of MATH-2 takes place by embryonic day 11.5 and initially localizes to the wall of brain vesicles and in the spinal cord. It is expressed in the cortical plate and the mantle layer in the developing central nervous system, and is limited to the nervous system in adults. Adult mouse cerebrums produce a high level of MATH-2 RNA with lower levels in other neuronal tissues. Research studies suggest that MATH-2 may function as a trans-acting factor involved in the development and maintenance of the mammalian nervous system.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MATH2/NEUROD6
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge