Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NAV2/RAINB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11892R-HRP
BOSSBS-11892R-HRPEA 900 EUR InStock
BOSSBS-11892R-HRP
Anti-NAV2/RAINB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Neuron navigator 2 (NAV2), also known as RAINB1 (retinoic acid inducible in neuroblastoma 1), POMFIL2 (pore membrane and/or filament-interacting-like protein 2), HELAD1 (helicase APC down-regulated 1), unc53H2 or steerin-2, is a 2,488 amino acid nuclear protein that plays a role in the development of sensory organs. Existing as thirteen alternatively spliced isoforms, Neuron navigator 2 displays 3' to 5' helicase activity and exonuclease activity and participates in the catalysis of ATP to ADP. Neuron navigator 2 is highly expressed in the nervous system of developing embryos and colon carcinomas, as well as in liver, brain and kidney. Lower levels of expression are found in lung, thyroid, spinal cord, heart, placenta and mammary gland. Neuron navigator 2 belongs to the Nav/unc-53 family and contains one CH (calponin-homology) domain.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: NAV2/RAINB1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge