Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NELL2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11895R-A488
BOSSBS-11895R-A488EA 893 EUR InStock
BOSSBS-11895R-A488
Anti-NELL2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
NELL2 is a secreted glycoprotein with one N-terminal TSP-like domain, five VWFC (von Willebrand factor C) domains and six EGF-like repeats that participate in calcium binding. NELL2 exists as a homotrimer associated with the endoplasmic reticulum (ER) and is specifically expressed in neurons playing a role in calcium-dependent intracellular events. NELL2 may act as a trophic factor in addition to its role as a signaling molecule implicated in the regulation of cell growth and differentiation. Due to alternative splicing, an additional NELL2 isoform exists that is identical to full length NELL2 except that it lacks the signal peptide that directs secretion. The additional isoform is therefore retained in the cytosol and functions as a non-secreted cytoplasmic protein that interacts with PKC bI.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NELL2/NRP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge