Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NHLH1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11901R-A350
BOSSBS-11901R-A350EA 893 EUR InStock
BOSSBS-11901R-A350
Anti-NHLH1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The helix-loop-helix (HLH) structures are known motifs commonly found in membrane-active and DNA-binding proteins. The helix-loop-helix proteins HEN1 and HEN2 are DNA-binding proteins that may be involved in cell-type determination in the early nervous system. Studies of expression in normal tissues have demonstrated expression of NHLH1/NSCL-1 and NHLH2/NSCL-2, the genes encoding HEN1 and HEN2, in the developing central and peripheral nervous system, specifically in developing neurons.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NHLH1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge