Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RAB3GAP2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11939R-FITC
BOSSBS-11939R-FITCEA 900 EUR InStock
BOSSBS-11939R-FITC
Anti-RAB3GAP2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
ab 3 GAP p150 is a ubiquitously expressed protein that contains 1,393 amino acids and belongs to the Rab3-GAP regulatory subunit family. Defects in Rab 3 GAP p150 are the cause of Martsolf and Warburg Micro syndrome. Both syndromes are characterized by congenital cataracts, microphthalmia, postnatal microcephaly and developmental delay, and are inherited in an autosomal recessive manner. The Rab3 GTPase-activating complex is a heterodimer composed of RAB3GAP and Rab 3 GAP p150 that interacts with DMXL2. Existing as two alternatively spliced isoforms, the Rab 3 GAP p150 gene is conserved in chimpanzee, dog, cow, rat, chicken, zebrafish, fruit fly, mosquito, A.thaliana and rice. The Rab 3 GAP p150 gene contains 36 exons and maps to human chromosome 1q41.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAB3GAP2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge