Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RAI1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11940R-CY5
BOSSBS-11940R-CY5EA 893 EUR InStock
BOSSBS-11940R-CY5
Anti-RAI1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Retinoic acid induced 1 (RAI1) is a 1,906 amino acid protein containing an N-terminal polyglutamine stretch that is expressed in most tissues, with highest expression in neuronal tissues. RAI1 functions as a transcriptional regulator and is important for embryonic and postnatal developments. Heterozygous deletions of the RAI1 gene are associated with Smith-Magenis syndrome (SMS), a mental retardation syndrome with behavioral, neurological and skeletal anomalies. Individuals affected with SMS usually display self-injurious behaviors, sleep disturbance, developmental delay and reduced motor and cognitive skills. RAI1 haploinsufficiency is specifically responsible for the obesity and craniofacial symptoms of SMS. RAI1 mutations have also been implicated in schizophrenia and spinocerebellar ataxia type 2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAI1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge