Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RPGRIP1L Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11946R-A647
BOSSBS-11946R-A647EA 893 EUR InStock
BOSSBS-11946R-A647
Anti-RPGRIP1L Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
RPGRIP1L is a 1,315 amino acid protein that belongs to the RPGRIP1 family and is thought to function in programmed cell death, craniofacial development and formation of the left-right axis. Existing as two alternatively spliced isoforms that localize to the cytoplasm, cytoskeleton, centrosome and cilium basal body, RPGRIP1L interacts with nephrocystin-4 and is moderately expressed in brain, retina and kidney. Containing two C2 domains, RPGRIP1L is encoded by a gene that maps to human chromosome 16q12.2. Defects in the gene encoding RPGRIP1L are the cause of Joubert syndrome type 7 (JBTS7), COACH syndrome (COACHS) and Meckel syndrome type 5 (MKS5).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RPGRIP1L
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge