Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SLITRK3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11956R-A680
BOSSBS-11956R-A680EA 890 EUR InStock
BOSSBS-11956R-A680
Anti-SLITRK3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The leucine-rich (LRR) repeat is a 20 to 30 amino acid motif that forms a hydrophobic å/ horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. SLITRK3 (SLIT and NTRK-like family, member 3) is a 977 amino acid single-pass type I membrane protein that contains 20 LRR repeats and belongs to the SLITRK family. Expressed at highest levels in cerebral cortex, SLITRK3 is also found in adult and fetal neural tissues and some astrocytic brain tumours. SLITRK3 functions to suppress neurite outgrowth and plays a role in the regulation of neuronal function. SLITRK3 is encoded by a gene that maps to human chromosome 3, which houses over 1100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci.

Type: Primary
Antigen: SLITRK3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 61-160/977
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge