Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SLITRK4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11957R-A488
BOSSBS-11957R-A488EA 893 EUR InStock
BOSSBS-11957R-A488
Anti-SLITRK4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
SLITRK family proteins are integral membrane proteins that have a C-terminal domain that is partially similar to TRK neurotrophin receptor proteins and two leucine-rich repeat (LRR) domains that are similar to those of SLIT proteins. SLITRK4 (SLIT and NTRK-like protein 4) is a 837 amino acid single-pass type I membrane protein that contains 18 LRR (leucine-rich) repeats and is expressed in neural tissues, specifically in the thalamus, hypothalamus, subventricular zone, CA3 region of the hippocampus and cortical plate. SLITRK4 may be upregulated in some astrocytic brain tumors such as glioblastomas, astrocytomas and primitive neuroectodermal tumors. As compared with its family member SLITRK2, SLITRK4 only weakly suppresses neurite outgrowth. A study using genome-wide transcriptional profiling suggested that the gene encoding SLITRK4, as well as the ARL5B and PLA2G7 genes, may be involved in the pathogenesis of preeclampsia.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLITRK4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge