Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SLITRK6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11960R-A750
BOSSBS-11960R-A750EA 890 EUR InStock
BOSSBS-11960R-A750
Anti-SLITRK6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The leucine-rich (LRR) repeat is a 20 to 30 amino acid motif that forms a hydrophobic ?/ horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. SLITRK6 (SLIT and NTRK-like family, member 6) is an 841 amino acid single-pass type I membrane protein that contains 16 LRR repeats and belongs to the SLITRK family. Expressed at high levels in select brain tissue, as well as in both adult and fetal lung, SLITRK6 functions to suppress neurite outgrowth, playing a role in the regulation of neuronal function. Multiple isoforms of SLITRK6 exist due to alternative splicing events.

Type: Primary
Antigen: SLITRK6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/841
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge