Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SORCS2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11963R-CY3
BOSSBS-11963R-CY3EA 900 EUR InStock
BOSSBS-11963R-CY3
Anti-SORCS2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
There are three SorCS genes that have diverse, partially overlapping functions in the central nervous system. In the developing and mature central nervous system, SorCS1, SorCS2 and SorCS3 genes are expressed in a combinatorial, non-overlapping pattern. SorCS proteins show homology to the mosaic receptor SorLA and the neurotensin receptor sortilin, based on a common VPS10 domain, which is the hallmark of the SorCS receptor family. SorCS2 (sortilin-related VPS10 domain containing receptor 2) is a 1,150 amino acid single-pass type I membrane protein that is highly expressed in brain and kidney. Containing six BNR repeats and a single PKD domain, SorCS2 is encoded by a gene that maps to human chromosome 4, which encodes nearly 6% of the human genome and has the largest gene deserts (regions of the genome with no protein encoding genes) of all of the human chromosomes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SORCS2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge