Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SRPX2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11967R-A488
BOSSBS-11967R-A488EA 893 EUR InStock
BOSSBS-11967R-A488
Anti-SRPX2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
SRPX2 is a 465 amino acid secreted protein expressed in neurons of the brain, including the rolandic area. It has been suggested that SRPX2 enhances cell motility, migration and adhesion through FAK signaling in gastric and other cancer cells. Localized to the cytoplasm, SRPX2 is a ligand for uPAR (urokinase plasminogen activator), a receptor that is a crucial component of the extracellular plasminogen proteolysis system. SRPX2 may be responsible for rolandic seizures (RSs) associated with oral and speech dyspraxia and mental retardation (MR). The involvement of SRPX2 in these disorders suggests an important role for SRPX2 in the perisylvian region critical for language and cognitive development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SRPX2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge