Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CACYBP Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11975R-CY5
BOSSBS-11975R-CY5EA 900 EUR InStock
BOSSBS-11975R-CY5
Anti-CACYBP Rabbit Polyclonal Antibody (Cy5®)
Antikörper
CacyBP is a 228 amino acid protein encoded by the human gene CACYBP. CacyBP is primarily a nuclear protein that contains one CS domain and one SGS domain. CacyBP is believed to be involved in calcium-dependent ubiquitination and subsequent proteosomal degradation of target proteins. It most likely serves as a molecular bridge in ubiquitin E3 complexes. It also participates in the ubiquitin-mediated degradation of b-catenin. CacyBP is thought to be a potential inhibitor of cell growth and invasion in the gastric cancer cell through its effects on b-catenin protein expression and transcriptional activation of TCF/LEF.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CACYBP
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge