Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NCS1/Frequenin Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11977R-A750
BOSSBS-11977R-A750EA 890 EUR InStock
BOSSBS-11977R-A750
Anti-NCS1/Frequenin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
NCS-1 (for neuronal calcium sensor-1, also designated frequenin) belongs to a highly conserved family of EF-hand-containing Ca++-binding proteins expressed mainly in neurons. NCS-1 is localised to neuronal cell bodies and axons throughout the brain and spinal cord. It is also expressed in glial cells and in neuroendocrine bovine adrenal chromaffin and PC12 cells. NCS-1 is a regulatory protein involved in Ca++-dependent exocytosis of synaptic vesicles and dense core granules. NCS-1 also functions in the voltage-independent autocrine pathway that negatively regulates non-L-type Ca++ channels.

Type: Primary
Antigen: NCS1/Frequenin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-50/190
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge