Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CACNB4/L-type Ca++ CP_4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11983R-A680
BOSSBS-11983R-A680EA 890 EUR InStock
BOSSBS-11983R-A680
Anti-CACNB4/L-type Ca++ CP_4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Voltage-dependent calcium channels are essential for the release of neurotransmitters. L-type (long lasting current) voltage-dependent calcium channels are composed of four subunits: an Alpha1 subunit, a Beta subunit, a Beta subunit and an Alpha2 Gamma subunit. The Beta subunit is encoded by four genes, designated Beta1-Beta 4, all of which contribute to the diversity of calcium currents and are involved in membrane trafficking of the Beta subunit. L-type Ca++ CP Beta 4, also known as CACNB4 (Calcium channel voltage-dependent subunit beta 4), CACNLB4 or CAB4, is a 484 amino acid protein that contains one SH3 domain and is expressed in ovary, brain and smooth muscle. Functioning as one of the four components of the Beta subunit, L-type Ca++ CP Beta 4 increases the peak calcium current in voltage-dependent calcium channels, thereby shifting the voltage dependencies of activation and inactivation and controlling G protein inhibition and Beta membrane targeting. Two isoforms of L-type Ca++ CP Beta4 exist due to alternative splicing events.

Type: Primary
Antigen: CACNB4/L-type Ca++ CP_4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/520
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge