Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CAPON Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11987R-HRP
BOSSBS-11987R-HRPEA 900 EUR InStock
BOSSBS-11987R-HRP
Anti-CAPON Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
CAPON (carboxy-terminal PDZ ligand of nNOS) selectively binds within the 100 amino acid PDZ domain of the neuronal nitric oxide synthase (nNOS), but not to endothelial NOS or inducible NOS, and sequesters nNOS in the cytosol. Biosynthesis of the neurotransmitter nitric oxide (NO) requires the association of nNOS with various synaptic proteins, including syntrophin, postsynaptic density (PSD)95 and PSD93 through a scaffolding PDZ domain. These proteins facilitate the transport of nNOS to the plasma membrane, where it is catalytically activated by NMDA-receptor mediated calcium channels. The association of nNOS with PSD95 or PSD93 is regulated by CAPON. The carboxy terminus of CAPON binds to the PDZ domain, competes with PSD95 and PSD93 for binding to nNOS and in turn prevents the translocation and catalytic activation of nNOS.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CAPON
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge